Skip to Content
Merck

E1523

Anti-ERK5 (big-MAPK, BMK1) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.44
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ARR, WB
Citations:
9
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 97 kDa

species reactivity

mouse, human

technique(s)

microarray: suitable, western blot: 1:8,000 using NIH 3T3 mouse fibroblasts cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPK7(5598)
mouse ... Mapk7(23939)

General description

Extracellular signal-regulated kinase 5 (ERK5), also referred to as big-MAP kinase 1 (BMK1), is a protein that belongs to mitogen-activated protein kinases family and is expressed mainly in skeletal muscle, kidney, heart and placenta. ERK5 contains a distinct 400-amino acid C-terminal and loop-12 domain, which includes two proline-rich regions.

Immunogen

synthetic peptide corresponding to amino acids 789-802 of human ERK5.

Application

Anti-ERK5 (big-MAPK, BMK1) an0tibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Extracellular signal-regulated kinase 5 (ERK5) has a critical role in different cellular processes like differentiation, proliferation and survival. Anti-ERK5 (big-MAPK, BMK1) antibody can be used to study the differential tissue expression and intracellular localization of the MAP Kinase isoform in normal and neoplastic tissues.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library