Skip to Content
Merck
All Photos(1)

Documents

E1378

Sigma-Aldrich

Escin

≥95%, powder

Synonym(s):

Aescin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

plant (Aesculus chinensis)

Quality Level

Assay

≥95%

form

powder

impurities

≤6.0% Water (Karl Fischer)

color

white

solubility

methanol: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

SMILES string

C\C=C(/C)C(=O)OC1[C@H](OC(C)=O)C2(CO)[C@H](O)C[C@]3(C)C(=CCC4C5(C)CCC(O[C@@H]6O[C@@H]([C@@H](O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)[C@H](O)[C@H]6O[C@H]8O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O)C(O)=O)[C@](C)(CO)C5CCC34C)C2CC1(C)C

InChI

1S/C55H86O24/c1-10-23(2)46(71)79-43-44(72-24(3)60)55(22-59)26(17-50(43,4)5)25-11-12-30-51(6)15-14-32(52(7,21-58)29(51)13-16-53(30,8)54(25,9)18-31(55)61)75-49-41(77-48-38(67)36(65)34(63)28(20-57)74-48)39(68)40(42(78-49)45(69)70)76-47-37(66)35(64)33(62)27(19-56)73-47/h10-11,26-44,47-49,56-59,61-68H,12-22H2,1-9H3,(H,69,70)/b23-10+/t26?,27-,28-,29?,30?,31-,32?,33-,34-,35+,36+,37-,38-,39+,40+,41-,42+,43?,44+,47+,48-,49-,51?,52-,53?,54-,55?/m1/s1

InChI key

AXNVHPCVMSNXNP-YSYFQUGBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Escin, a natural mixture of triterpenoid saponins isolated from horse chestnut (Aesculus hippocastanum) seeds, is used and studied as a vasoprotective anti-inflammatory, anti-edematous and anti-nociceptive agent. It may be used to study its pharmacokinetics, safety and efficacy as a chemosensitizer and modulator of NF-κ B cell signaling.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenyu Xin et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 18(4), 272-277 (2010-09-21)
Escin, a natural mixture of triterpenoid saponins isolated from the seed of the horse chestnut (Aesculus hippocastanum), had been demonstrated to possess anti-edematous and anti-inflammatory effects. The present study was designed to investigate whether escin exhibits synergistic anti-inflammatory effects when
Kuzhuvelil B Harikumar et al.
Molecular pharmacology, 77(5), 818-827 (2010-01-28)
Agents that can enhance tumor cell apoptosis and inhibit invasion have potential for the treatment of cancer. Here, we report the identification of escin, a pentacyclic triterpenoid from horse chestnut that exhibits antitumor potential against leukemia and multiple myeloma. Whether
Hang Zhao et al.
Canadian journal of anaesthesia = Journal canadien d'anesthesie, 54(3), 201-207 (2007-03-03)
The purpose of this study was to determine if intrathecal landiolol, a beta1-blocker, can modulate formalin-induced nociception and spinal c-Fos expression in mice, in the absence of anesthesia. Thirty-two mice were randomly assigned to one of four groups: the control
J S Fan et al.
Pflugers Archiv : European journal of physiology, 436(6), 1021-1023 (1998-11-03)
Perforated patch recording with nystatin, amphotericin B and gramicidin can be more difficult in the hands of some investigators than others. In addition, it is difficult to introduce low molecular weight substances such as dyes into the cytoplasm in such
Xiu-Jun Wu et al.
Journal of ethnopharmacology, 151(2), 839-845 (2013-12-18)
Adequate pharmacokinetic data of escin, a natural mixture of triterpene saponins used for the treatment of chronic venous insufficiency, hemorrhoids, inflammation and edema, is of special interest in view of the growing use of escin agent in clinical medicine. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service