Skip to Content
Merck

Skip To

E0781

Extract-N-Amp PCR ReadyMix with qPCR Reference dye

125 mL sufficient for 10000 amplifications, hotstart, dNTPs included

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.55
UNSPSC Code:
41106303
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Amplification ReadyMix to supplement Extract-N-Amp Plant and Extract-N-Amp Tissue kits

Quality Segment

usage

sufficient for 10000 reactions, 125 mL sufficient for 10000 amplifications

feature

dNTPs included, hotstart

application(s)

agriculture

detection method

probe-based

shipped in

wet ice

storage temp.

−20°C

General description

Extract-N-Amp PCR ReadyMix with qPCR Reference dye is intended for use with Sigma′s Extract-N-Amp Plant kit and and Extract-N-Amp Tissue PCR kit. All kits include a PCR ReadyMix sufficient for one PCR reaction per extraction. However, if additional PCR reactions are required, supplemental PCR ReadyMix may be needed. Sigma′s Reference Dye for qPCR is a proprietary dye for use with real-time PCR. It is used for normalization of reaction data when using SYBR Green, molecular probes, or dual-labeled probe chemistries for real-time detection. Instrument settings for ROX reference dye are satisfactory for the measurement of Reference Dye for qPCR.

Legal Information

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P8115R4775L6669
application(s)

agriculture

application(s)

-

application(s)

agriculture

application(s)

agriculture

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

usage

sufficient for 10000 reactions, 125 mL sufficient for 10000 amplifications

usage

sufficient for 1000 reactions, 12 mL sufficient for 1000 amplifications

usage

sufficient for 100 reactions, 1.2 mL sufficient for 100 amplifications, sufficient for 1000 reactions, 12 mL sufficient for 1000 amplifications, sufficient for 10000 reactions, 125 mL sufficient for 10000 amplifications

usage

sufficient for 100 reactions, sufficient for 2000 reactions, sufficient for 500 reactions

feature

hotstart, dNTPs included

feature

dNTPs included, hotstart

feature

dNTPs included, hotstart

feature

dNTPs included, hotstart


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

wgk

WGK 2

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters