Skip to Content
Merck
All Photos(3)

Key Documents

D9758

Sigma-Aldrich

Dilaurin Mixed Isomers

≥99%

Synonym(s):

Dilaurin Isomer Mixture

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H52O5
CAS Number:
Molecular Weight:
456.70
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

plant kernels (palm)

Assay

≥99%

form

powder

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCC(=O)OCC(O)COC(=O)CCCCCCCCCCC.CCCCCCCCCCCC(=O)OCC(CO)OC(=O)CCCCCCCCCCC

InChI

1S/2C27H52O5/c1-3-5-7-9-11-13-15-17-19-21-26(29)31-23-25(28)24-32-27(30)22-20-18-16-14-12-10-8-6-4-2;1-3-5-7-9-11-13-15-17-19-21-26(29)31-24-25(23-28)32-27(30)22-20-18-16-14-12-10-8-6-4-2/h2*25,28H,3-24H2,1-2H3

InChI key

RURSPNRVQHSFCU-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycerol conformation and molecular packing of membrane lipids. The crystal structure of 2,3-dilauroyl-D-glycerol.
I Pascher et al.
Journal of molecular biology, 153(3), 791-806 (1981-12-15)
G Vanderkooi
Chemistry and physics of lipids, 55(3), 253-264 (1990-09-01)
Computational procedures have been developed by which the total energy of a lipid multibilayer can be calculated and minimized. The energy is expressed as a sum of non-bonded, electrostatic, hydrogen bonded and torsional energy terms and includes intramolecular and intermolecular
D Lairon et al.
Biochimica et biophysica acta, 618(1), 106-118 (1980-04-18)
We report a new technique which allows us to follow the lipolysis of monomolecular films in the presence of bile salts by using a 'zero-order' trough (Verger, R. and de Haas, G.H. (1973) Chem. Phys. Lipids 10, 127). The effects
C M Mansbach et al.
The Journal of clinical investigation, 69(2), 368-376 (1982-02-01)
We evaluated phospholipase activity in the intestine of rats and other species. Phospholipase activity was assayed by a surface barostat technique or an egg yolk titration system. Mucosal activity was found only by the surface barostat technique with phosphatidylglycerol as
J A Hamilton et al.
The Journal of biological chemistry, 266(2), 1177-1186 (1991-01-15)
The interaction of diacylglycerols, primarily 1,2-dilauroyl-sn-glycerol (1,2-DLG), with egg phosphatidylcholine (PC) bilayers was studied by NMR spectroscopy and other physical techniques. In the low proportions used (less than or equal to 20 mol % with respect to total lipid), 1,2-DLG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service