Skip to Content
Merck

D9134

2,3-Diphospho-D-glyceric acid penta(cyclohexylammonium) salt

≥97% (TLC)

Synonym(s):

D-Glycerate 2,3-diphosphate pentacyclohexylamine salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3H8O10P2 · 5C6H13N
CAS Number:
Molecular Weight:
761.91
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

egg white

Quality Segment

assay

≥97% (TLC)

form

powder

impurities

≤1% 3-phosphoglyceric acid

color

white

solubility

H2O: 50 mg/mL, clear, colorless to very faintly yellow

storage temp.

−20°C

SMILES string

NC1CCCCC1.NC2CCCCC2.NC3CCCCC3.NC4CCCCC4.NC5CCCCC5.OC(=O)[C@@H](COP(O)(O)=O)OP(O)(O)=O

InChI

1S/5C6H13N.C3H8O10P2/c5*7-6-4-2-1-3-5-6;4-3(5)2(13-15(9,10)11)1-12-14(6,7)8/h5*6H,1-5,7H2;2H,1H2,(H,4,5)(H2,6,7,8)(H2,9,10,11)/t;;;;;2-/m.....1/s1

InChI key

WMBJYTVZNODJKS-SPUYNGLASA-N

Application

D-Glycerate 2,3-diphosphate (DPG), cofactor of both phosphoglyceric acid mutase and hemoglobin, may be used as a reference compound in analysis of blood cell (erythrocyte) glycolysic cycle metabolites. 2,3-diphosphoglycerate (DPG) may be used as a reference in assays that measure DPG levels to indicate the health status of erythrocytes/red blood cells (RBC).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library