Skip to Content
Merck

D8768

cis-4,7,10,13,16,19-Docosahexaenoic acid sodium salt

≥95%, waxy solid

Synonym(s):

Sodium (all-Z)-4,7,10,13,16,19-docosahexaenoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C22H31O2Na
CAS Number:
Molecular Weight:
350.47
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

cod liver oil

Quality Segment

assay

≥95%

form

waxy solid

functional group

carboxylic acid

lipid type

omega FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCC(O[Na])=O

InChI

1S/C22H32O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24;/h3-4,6-7,9-10,12-13,15-16,18-19H,2,5,8,11,14,17,20-21H2,1H3,(H,23,24);/q;+1/p-1/b4-3-,7-6-,10-9-,13-12-,16-15-,19-18-;

InChI key

SNNDEWVSGZRIFE-FPYKSTABSA-M



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library