Skip to Content
Merck
All Photos(3)

Key Documents

D8040

Sigma-Aldrich

(±)-trans-U-50488 methanesulfonate salt

Synonym(s):

trans-(±)-3,4-Dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzeneacetamide methanesulfonate salt, U-50488H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26Cl2N2O · CH4O3S
Molecular Weight:
465.43
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

solubility

H2O: 50 mg/mL

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.CN([C@@H]1CCCC[C@H]1N2CCCC2)C(=O)Cc3ccc(Cl)c(Cl)c3

InChI

1S/C19H26Cl2N2O.CH4O3S/c1-22(19(24)13-14-8-9-15(20)16(21)12-14)17-6-2-3-7-18(17)23-10-4-5-11-23;1-5(2,3)4/h8-9,12,17-18H,2-7,10-11,13H2,1H3;1H3,(H,2,3,4)/t17-,18-;/m1./s1

InChI key

OJPHNZCUXUUVKU-JAXOOIEVSA-N

Gene Information

human ... OPRK1(4986)

Application

(±)-trans-U-50488 methanesulfonate salt has been used as a control agonist for κ-opioid receptor.

Biochem/physiol Actions

Selective κ-opioid receptor agonist. (±)-trans-U-50488 methanesulfonate salt is known to induce diuresis. U-50488H is found to prevent morphine tolerance when administered along with it.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Opioid receptor delta as a global modulator of skin differentiation and barrier function repair
Chajra H, et al.
International Journal of Cosmetic Science, 37(4), 386-394 (2015)
Guang-Jie Gao et al.
Evidence-based complementary and alternative medicine : eCAM, 2022, 2868135-2868135 (2022-08-30)
Acute lung injury (ALI) is one of the common and severe complications of cardiopulmonary bypass (CPB), which is the primary cause of death in intensive care units. Nevertheless, there is a lack of effective treatment for ALI secondary to CPB.
Differential effects of the novel kappa opioid receptor antagonist, JDTic, on reinstatement of cocaine-seeking induced by footshock stressors vs cocaine primes and its antidepressant-like effects in rats
Beardsley PM, et al.
Psychopharmacology, 183(1), 118-118 (2005)
Alena Karnošová et al.
International journal of molecular sciences, 22(16) (2021-08-28)
The anorexigenic neuropeptide prolactin-releasing peptide (PrRP) is involved in the regulation of food intake and energy expenditure. Lipidization of PrRP stabilizes the peptide, facilitates central effect after peripheral administration and increases its affinity for its receptor, GPR10, and for the
X C Yu et al.
Journal of molecular and cellular cardiology, 31(10), 1809-1819 (1999-10-20)
During myocardial ischaemia the beta-adrenoceptor is activated, which contributes, at least partly, to cardiac arrhythmias via inducing [Ca2+]i oscillations. Since beta-adrenoceptor is negatively modulated by the kappa-opioid receptor in the heart, the present study attempted to determine if kappa-opioid receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service