Skip to Content
Merck
All Photos(1)

Key Documents

D7017

Sigma-Aldrich

Dynorphin A Porcine Fragment 1-13

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C75H126N24O15
CAS Number:
Molecular Weight:
1603.95
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97% (HPLC)

form

powder

composition

Peptide content, ≥50%

UniProt accession no.

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(CCCNC(N)=N)NC(=O)C(CCCNC(N)=N)NC(=O)C(CC(C)C)NC(=O)C(Cc1ccccc1)NC(=O)CNC(=O)CNC(=O)C(N)Cc2ccc(O)cc2)C(=O)NC(CCCNC(N)=N)C(=O)N3CCCC3C(=O)NC(CCCCN)C(=O)NC(CC(C)C)C(=O)NC(CCCCN)C(O)=O

InChI

1S/C75H126N24O15/c1-7-45(6)61(70(111)94-53(25-17-35-87-75(83)84)71(112)99-36-18-26-58(99)69(110)93-50(21-11-13-31-76)64(105)96-56(38-44(4)5)67(108)95-54(72(113)114)22-12-14-32-77)98-65(106)52(24-16-34-86-74(81)82)91-63(104)51(23-15-33-85-73(79)80)92-66(107)55(37-43(2)3)97-68(109)57(40-46-19-9-8-10-20-46)90-60(102)42-88-59(101)41-89-62(103)49(78)39-47-27-29-48(100)30-28-47/h8-10,19-20,27-30,43-45,49-58,61,100H,7,11-18,21-26,31-42,76-78H2,1-6H3,(H,88,101)(H,89,103)(H,90,102)(H,91,104)(H,92,107)(H,93,110)(H,94,111)(H,95,108)(H,96,105)(H,97,109)(H,98,106)(H,113,114)(H4,79,80,85)(H4,81,82,86)(H4,83,84,87)

InChI key

OVVIBUHLQIYUEU-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Ile-Arg-Pro-Lys-Leu-Lys

Application

Dynorphin A Porcine Fragment 1-13 has been used as a test compound in the field asymmetric waveform ion mobility spectrometry coupled to mass spectrometry (FAIMS/IMS/MS).

Biochem/physiol Actions

Dynorphin (Dyn) (1-13), a bioactive product of dynorphin A displays interaction with N-methyl- D-aspartate (NMDA) as well as with opioid receptors. It elevates intracellular calcium [Ca2+]i level and contributes to the loss of neurons affecting neuronal viability.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evgeniy S Petrov et al.
Behavioural brain research, 170(1), 1-14 (2006-04-20)
The role of endogenous opioid system in learning (Pavlovian conditioning) and reinforcement was studied in newborn rats by pairing central injection of dynorphin A (1-13), an endogenous ligand with high affinity for kappa opioid receptors, with a surrogate nipple. For
Yue'e Peng et al.
Analytical chemistry, 83(23), 8863-8866 (2011-10-29)
A novel ionization device for controlling the charge states of peptides based on an inductive elecrospray ionization technique was developed. This ion source keeps the major capabilities of electrospray ionization (ESI) which is compatible with liquid separation techniques (such as
Jun Yang et al.
Neuroscience research, 54(1), 49-56 (2005-11-29)
Our previous study proved that the hypothalamic paraventricular nucleus (PVH) plays an important role in acupuncture analgesia. The neuropeptides involving in the PVH regulation of acupuncture analgesia was investigated in the rat. The changes of pain threshold, which was induced
Michael E Nizhnikov et al.
Alcohol (Fayetteville, N.Y.), 41(7), 525-534 (2007-11-06)
Recent studies have systematically indicated that newborn rats are highly sensitive to ethanol's positive reinforcing effects. Central administrations of ethanol (25-200mg %) associated with an olfactory conditioned stimulus (CS) promote subsequent conditioned approach to the CS as evaluated through the
M Ogura et al.
Journal of neurophysiology, 83(6), 3366-3376 (2000-06-10)
The opioids contained in striato-pallidal axons are thought to play a significant role in motor control. We examined post- and presynaptic effects of the kappa (kappa)-receptor agonist dynorphin A (1-13) (DYN13) on the globus pallidus (GP) neurons in rat brain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service