Skip to Content
Merck
All Photos(1)

Documents

D4921

Sigma-Aldrich

Destruxin A from Metarrhizium anisopliae

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H47N5O7
CAS Number:
Molecular Weight:
577.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

biological source

Metarrhizium anisopliae

Quality Level

Assay

≥98% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@@H]1NC(=O)[C@H]2CCCN2C(=O)[C@H](CC=C)OC(=O)CCNC(=O)[C@H](C)N(C)C(=O)[C@H](C(C)C)N(C)C1=O

InChI

1S/C29H47N5O7/c1-9-12-21-27(38)34-16-11-13-20(34)26(37)31-23(18(5)10-2)28(39)33(8)24(17(3)4)29(40)32(7)19(6)25(36)30-15-14-22(35)41-21/h9,17-21,23-24H,1,10-16H2,2-8H3,(H,30,36)(H,31,37)/t18-,19-,20+,21-,23-,24-/m0/s1

InChI key

XIYSEKITPHTMJT-UTJJDKHZSA-N

Application

Destruxin A from Metarrhizium anisopliae has been used for immersion and inoculation bioassay in Rhipicephalus (Boophilus) microplus. It has been used as a transport inhibitor in Drosophila melanogaster.

Biochem/physiol Actions

Destruxin A is a cyclic hexadepsipeptide produced by fungus causing paralysis and death in insects. May suppress the insect immune response. Possesses an inhibitory activity on leukemic cell proliferation, decreasing the number of cells in G2/M phase. It is virustatic against arboviruses.

Quality

Balance is other destruxins

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Hutwimmer et al.
Mycologia, 102(1), 1-10 (2010-02-03)
Nutritional conditions causing droplet exudation by Metarhizium anisopliae var. anisopliae were studied. Exudation in droplets occurred only on media with more than one carbon source and was highly dependent on the ratio of a well metabolized sugar such as trehalose
Esau Ruiz-Sanchez et al.
Toxicon : official journal of the International Society on Toxinology, 56(6), 1043-1051 (2010-07-31)
Destruxins, a family of cyclic peptides, are produced by various species of entomopathogenic fungi. These peptides have been shown to influence calcium-dependent processes in insect cell lines and tissues, such as skeletal muscles. To better understand the mechanism of action
Xiang Meng et al.
Toxicon : official journal of the International Society on Toxinology, 58(4), 327-335 (2011-07-02)
The cytotoxicity of a destruxin A (DA) treatment of Spodoptera litura SL-1 cells was investigated. An MTT assay showed that DA was highly toxic to SL-1 cells in a concentration- and time-dependent manner. The IC(50) values of DA, after 24 h
Esau Ruiz-Sanchez et al.
Toxicon : official journal of the International Society on Toxinology, 55(6), 1162-1170 (2010-01-12)
The production of peptide toxins by entomopathogenic fungi during the infection process plays critical roles in pathogenesis. To gain insight into the mechanism of action of these mycotoxins on insect internal organs, we have evaluated the effects of destruxin A
Esau Ruiz-Sanchez et al.
Archives of insect biochemistry and physiology, 80(2), 109-122 (2012-04-24)
Destruxins have been implicated in the infection process by entomopathogenic fungi and have been also found to be highly toxic when applied topically or ingested by different insect species. To gain insight into the mechanism of action of this toxin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service