Skip to Content

D4630

2,5-Diphenyloxazole

suitable for liquid scintillation spectrometry

Synonym(s):

DPO, PPO

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H11NO
CAS Number:
Molecular Weight:
221.25
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
202-181-3
Beilstein/REAXYS Number:
157021
MDL number:
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

bp

360 °C (lit.)

mp

72-74 °C (lit.)

suitability

suitable for liquid scintillation spectrometry

functional group

phenyl

SMILES string

c1ccc(cc1)-c2cnc(o2)-c3ccccc3

InChI

1S/C15H11NO/c1-3-7-12(8-4-1)14-11-16-15(17-14)13-9-5-2-6-10-13/h1-11H

InChI key

CNRNYORZJGVOSY-UHFFFAOYSA-N

General description

2,5-Diphenyloxazole (also known as PPO or p-phenylphenoxazole) is a fluorescent organic compound. It can be also used as an organic scintillator due to its high photoluminescence quantum yield and intriguing luminescent properties. It is widely used as a fluorescent probe or dye in analytical chemistry and biochemistry.

Application

2,5-Diphenyloxazole (PPO) can be used in:
  • fluorography techniques for the detection of tritium, tritiated glycopeptides, and 35S labeled proteins.
  • liquid scintillation cocktails as primary and secondary fluors.


Still not finding the right product?

Explore all of our products under 2,5-Diphenyloxazole


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

386.6 - 393.8 °F - closed cup

flash_point_c

197 - 201 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library