Skip to Content
Merck
All Photos(1)

Key Documents

D4268

Sigma-Aldrich

NG,NG-Dimethylarginine dihydrochloride

≥98% (TLC), powder, NOS inhibitor

Synonym(s):

asym-Dimethylarginine, ADMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N4O2 · 2HCl
CAS Number:
Molecular Weight:
275.18
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

Product Name

NG,NG-Dimethylarginine dihydrochloride,

Assay

≥98% (TLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

Cl[H].Cl[H].CN(C)C(=N)NCCC[C@H](N)C(O)=O

InChI

1S/C8H18N4O2.2ClH/c1-12(2)8(10)11-5-3-4-6(9)7(13)14;;/h6H,3-5,9H2,1-2H3,(H2,10,11)(H,13,14);2*1H/t6-;;/m0../s1

InChI key

SYLNVYJOPZWPJI-ILKKLZGPSA-N

General description

NG,NG-Dimethylarginine dihydrochloride is a methylated arginine derivative, which is produced by the liver. It is linked to liver cirrhosis, alcoholic hepatitis and acute liver failure (ALF).

Application

NG,NG- Dimethylarginine dihydrochloride has been used:
  • to study its effects on cardiovascular activities in rostral ventrolateral medulla (RVLM) in rats
  • as a standard for measuring L-arginine and dimethylarginines
  • to study its effects on hepatic deterioration

Biochem/physiol Actions

Reversible inhibitor of nitric oxide synthetase in vivo and in vitro.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Rodling Wahlström et al.
SpringerPlus, 3, 98-98 (2014-03-07)
A prospective, randomised, double-blinded, clinical trial was performed at a level 1 trauma centre to determine if a prostacyclin analogue, epoprostenol (Flolan®), could attenuate systemic inflammatory response in patients with severe traumatic brain injury (TBI). 46 patients with severe TBI
Kyoung Min Kim et al.
Cardiovascular diabetology, 16(1), 146-146 (2017-11-11)
The cardiovascular benefits of statins have been proven, but their effect on circulation in small vessels has not been examined fully. We investigated the effect of 20 mg rosuvastatin on biomarkers, including paraoxonase-1 (PON-1) and asymmetric dimethylarginine (ADMA), and on microvascular
Aleksandar Radujkovic et al.
Haematologica, 104(4), 827-834 (2018-12-06)
Increasing evidence suggests that endothelial cell distress is associated with mortality after allogeneic stem cell transplantation and acute graft-versus-host disease. Asymmetric dimethylarginine is an endogenous nitric oxide synthase inhibitor that induces endothelial cell dysfunction. We analyzed the impact of pre-transplant
José D Martins et al.
Journal of the American Heart Association, 8(7), e011536-e011536 (2019-04-02)
Background Optimally treated patients with coarctation of the aorta remain at risk for late vascular dysfunction. The effect of treatment modality on vascular function is unknown. The LOVE-COARCT (Long-term Outcomes and Vascular Evaluation After Successful Coarctation of the Aorta Treatment)
P Vallance et al.
Lancet (London, England), 339(8793), 572-575 (1992-03-07)
Nitric oxide (NO), synthesised from L-arginine, contributes to the regulation of blood pressure and to host defence. We describe in-vitro and in-vivo evidence that NO synthesis can be inhibited by an endogenous compound, NG,NG-dimethylarginine (asymmetrical dimethylarginine, ADMA). In man, this

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service