Skip to Content
Merck

Skip To

D4138

Deoxyribonuclease II from porcine spleen

Type IV, lyophilized powder, 2,000-6,000 Kunitz units/mg protein (biuret)

Synonym(s):

DNase II, Deoxyribonucleate 3′-oligonucleotido-hydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-801-8
MDL number:
EC Number:
Specific activity:
2,000-6,000 Kunitz units/mg protein (biuret)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


type

Type IV

Quality Segment

form

lyophilized powder

specific activity

2,000-6,000 Kunitz units/mg protein (biuret)

mol wt

45 kDa

composition

Protein, 60-80%

input

general use sample(s)

foreign activity

RNase ≤0.05%

storage temp.

−20°C

Application

DNase II from Sigma has been used to treat transformed cells during the purification of β-lactamase. It has also been used for the preparation of adenoma tissue in a study that investigated the effect of somatoprim on growth hormone secretion in human adenoma cell cultures (hSA).
Deoxyribonuclease II from porcine spleen has been used in an immunohistological study of the immune system cells in paraffin-embedded tissues. Deoxyribonuclease II from porcine spleen has also been used in a study to investigate its reassociation with the lysosomal membrane.

Biochem/physiol Actions

The enzyme is a lysosomal acid hydrolase that hydrolyzes DNA to 3′-phosphoryl oligonucleotides in the absence of metal ions, under acidic conditions. It is a non-covalently linked α·β heterodimer of approximately 45,000 Da molecular mass.

Physical form

Contains sodium chloride

Other Notes

One Kunitz unit will produce a ΔA260 of 0.001 per min per mL at pH 4.6 at 25°C; [Mg2+] = 0.83 mM

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D8764D4513D4527
description

Type IV, lyophilized powder, 2,000-6,000 Kunitz units/mg protein (biuret)

description

Type V, essentially salt-free, lyophilized powder, ≥1,000 units/mg protein

description

Type II-S, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein

description

Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein

specific activity

2,000-6,000 Kunitz units/mg protein (biuret)

specific activity

≥1,000 units/mg protein

specific activity

≥2,000 units/mg protein

specific activity

≥2,000 units/mg protein

form

lyophilized powder

form

essentially salt-free, lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

mol wt

45 kDa

mol wt

38 kDa

mol wt

~31 kDa

mol wt

~31 kDa

foreign activity

RNase ≤0.05%

foreign activity

RNase ≤0.1%

foreign activity

Chymotrypsin ≤0.01%, Protease ≤0.005%, RNase ≤0.01%

foreign activity

Chymotrypsin ≤0.01%, Protease ≤0.005%, RNase ≤0.01%

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

300


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters