Skip to Content
Merck
All Photos(3)

Documents

D4022

Sigma-Aldrich

P1,P5-Di(adenosine-5′) pentaphosphate pentasodium salt

≥95% (HPLC), powder

Synonym(s):

A(5′)P5(5′)A, Diadenosine pentaphosphate pentasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H29N10O22P5 · 5 Na
CAS Number:
Molecular Weight:
1026.28
Beilstein:
4949216
EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥95% (HPLC)

form

powder

mol wt

~_1.0 kDa

color

white to yellow-white

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP([O-])(=O)OP([O-])(=O)OP([O-])(=O)OC[C@H]4O[C@H]([C@H](O)[C@@H]4O)n5cnc6c(N)ncnc56)[C@@H](O)[C@H]3O

InChI

1S/C20H29N10O22P5.5Na/c21-15-9-17(25-3-23-15)29(5-27-9)19-13(33)11(31)7(47-19)1-45-53(35,36)49-55(39,40)51-57(43,44)52-56(41,42)50-54(37,38)46-2-8-12(32)14(34)20(48-8)30-6-28-10-16(22)24-4-26-18(10)30;;;;;/h3-8,11-14,19-20,31-34H,1-2H2,(H,35,36)(H,37,38)(H,39,40)(H,41,42)(H,43,44)(H2,21,23,25)(H2,22,24,26);;;;;/q;5*+1/p-5/t7-,8-,11-,12-,13-,14-,19-,20-;;;;;/m1...../s1

InChI key

NNMFUJJMJIYTSP-CSMIRWGRSA-I

Looking for similar products? Visit Product Comparison Guide

General description

P1,P5-Di(adenosine-5′) pentaphosphate (Ap5A) is a diadenosine polyphosphate, which has a tail-to-tail dimer structure. Ap5A is synthesized in a twostep process involving the formation of adenosine 5′-tetraphosphate (P4A) from the ATP and trimeta-phosphate (P3). In the second step, P4A is converted to Ap5A. The synthesis of Ap5A requires a pH optimum in the range of 7.5 to 8.5 and is modulated by metal ions.

Application

P1,P5-Di(adenosine-5′) pentaphosphate pentasodium salt has been used as:
  • an adenylate kinase (AK) inhibitor in: sarcoma osteogenic (Saos-2) cells
  • mitochondrial lysates during ATP synthesis
  • tetramethylrhodamine methyl ester (TMRM) based membrane potential assay
  • chromoplasts

Biochem/physiol Actions

A diadenosine polyphosphate stored in secretory granules of thrombocytes, chromaffin and neuronal cells. After release into the extracellular space, it affects a variety of biological activities in a wide range of target tissues. In the nervous system it acts through various purinergic receptors. It also activates 5′-nucleotidase and inhibits adenosine kinase activity in vitro. Ap5A is metabolized by soluble enzymes in the blood plasma and by membrane-bound ectoenzymes of a number of cell types including endothelial and smooth muscle cells. In cardiac muscle, pM to nM concentrations significantly increase the open-probability of ryanodine-receptor (RyR2) gates, with prolonged action due to slow dissociation from the receptor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile and selective synthesis of diadenosine polyphosphates through catalysis by leucyl t-RNA synthetase coupled with ATP regeneration
Nakajima H, et al.
Agricultural and Biological Chemistry, 53(3), 615-623 (1989)
Isolation of Tomato Fruit Chromoplasts and Determination of ATP Levels
Yaakov B
The Plant Journal (2013)
Soluble adenylyl cyclase-mediated cAMP signaling and the putative role of PKA and EPAC in cerebral mitochondrial function
Jakobsen E, et al.
Journal of Neuroscience Research (2019)
Gourav Bhardwaj et al.
The Journal of clinical investigation, 131(18) (2021-08-04)
Decreased skeletal muscle strength and mitochondrial dysfunction are characteristic of diabetes. The actions of insulin and IGF-1 through the insulin receptor (IR) and IGF-1 receptor (IGF1R) maintain muscle mass via suppression of forkhead box O (FoxO) transcription factors, but whether
Polyphosphate as a donor of high-energy phosphate for the synthesis of ADP and ATP
Muller WEG, et al.
Journal of Cell Science, 130(16), 2747-2756 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service