Skip to Content
Merck

D2194

Anti-Derlin-3 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DERL3, Anti-Der1-like domain family, member 3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IP, WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~26 kDa

species reactivity

rat (predicted), human, mouse (predicted)

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using extract of HEK-293T cells expressing recombinant human Derlin-3, indirect immunofluorescence: 10-20 μg/mL using human HeLa cells, western blot: 2-5 μg/mL using whole extract of HEK-293T cells expressing recombinant human Derlin-3

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DERL3(91319)
mouse ... Derl3(70377)
rat ... Derl3(690315)

General description

Derlin-3 is a mammalian homologues of yeast Der1p, a transmembrane protein required for yeast endoplasmic reticulum (ER)-associated degradation (ERAD). Derlin 3 is encoded by the gene mapped to human chromosome 22q11.23. It is expressed in the ER and is a member of the derlin protein family.

Immunogen

synthetic peptide corresponding to amino acid residues 192-205 of human Derlin-3, conjugated to KLH. The corresponding sequence differs by 3 amino acids in mouse and 2 amino acids in rat.

Application

Anti-Derlin-3 antibody produced in rabbit has been used in immunoblotting, immunoprecipitation, and immunofluorescence.

Biochem/physiol Actions

Derlin-3 is involved in the degradation of misfolded glycoproteins in the ER. Derlin-3 shares ~30% sequence identity with Derlin-1 and spans the lipid bilayer of the ER four times, showing structural similarity to Derlin-1. It is a component of the mammalian ER-associated degradation (ERAD) mechanism and is upregulated by unfolded protein response (UPR). Overexpression of this gene leads to increase in degradation of misfolded glycoprotein, whereas its knockdown blocks degradation. Derlin-3 also interacts with the mammalian orthologs of the yeast Hrd1p/Hrd3p ubiquitin-ligase complex.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)

wgk

nwg

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library