Skip to Content
Merck

Skip To

D127

Dextrorphan tartrate

NMDA-receptor antagonist, powder

Synonym(s):

(+)-3-Hydroxy-N-methylmorphinan (+)-tartrate salt, Dextrorphan D-tartrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H23NO · C4H6O6
CAS Number:
Molecular Weight:
407.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Dextrorphan tartrate, powder

description

Drug Control: Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet

Quality Segment

form

powder

drug control

Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet

color

white

solubility

H2O: >10 mg/mL

SMILES string

O[C@@H]([C@H](O)C(O)=O)C(O)=O.CN1CC[C@@]23CCCC[C@@H]2[C@@H]1Cc4ccc(O)cc34

InChI

1S/C17H23NO.C4H6O6/c1-18-9-8-17-7-3-2-4-14(17)16(18)10-12-5-6-13(19)11-15(12)17;5-1(3(7)8)2(6)4(9)10/h5-6,11,14,16,19H,2-4,7-10H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t14-,16+,17+;1-,2-/m10/s1

InChI key

RWTWIZDKEIWLKQ-FEBOIHDFSA-N

Application

Dextrorphan tartrate has been used to study its effects on blood glucose levels by targeting pancreatic islets.[1]

Biochem/physiol Actions

Dextrorphan is a noncompetitive NMDA glutamate receptor antagonist that attenuates glutamate neurotoxicity in cortical cell cultures.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G1501PHR1974G1626
Quality Level

200

Quality Level

300

Quality Level

300

Quality Level

300

form

powder

form

powder

form

powder

form

powder

Gene Information

human ... GRIN1(2902), GRIN2A(2903), GRIN2B(2904), GRIN2C(2905), GRIN2D(2906), GRINA(2907)

Gene Information

human ... GRIK1(2897), GRIK2(2898), GRIK3(2899), GRIK4(2900), GRIN1(2902), GRIN2A(2903), GRIN2B(2904), GRIN2C(2905), GRIN2D(2906), GRIN3A(116443), GRIN3B(116444), GRINA(2907)
mouse ... GRIK1(14805), GRIK2(14806), GRIK3(14807), GRIK4(110637), GRIN1(14810), GRIN2A(14811), GRIN2B(14812), GRIN2C(14813), GRIN2D(14814), GRIN3A(242443), GRIN3B(170483), GRINA(66168)
rat ... GRIK1(29559), GRIK2(54257), GRIK3(298521), GRIK4(24406), GRIN1(24408), GRIN2A(24409), GRIN2B(24410), GRIN2C(24411), GRIN2D(24412), GRIN3A(191573), GRIN3B(170796), GRINA(266668)

Gene Information

-

Gene Information

human ... GRIK1(2897), GRIK2(2898), GRIK3(2899), GRIK4(2900), GRIN1(2902), GRIN2A(2903), GRIN2B(2904), GRIN2C(2905), GRIN2D(2906), GRIN3A(116443), GRIN3B(116444), GRINA(2907)
mouse ... GRIK1(14805), GRIK2(14806), GRIK3(14807), GRIK4(110637), GRIN1(14810), GRIN2A(14811), GRIN2B(14812), GRIN2C(14813), GRIN2D(14814), GRIN3A(242443), GRIN3B(170483), GRINA(66168)
rat ... GRIK1(29559), GRIK2(54257), GRIK3(298521), GRIK4(24406), GRIN1(24408), GRIN2A(24409), GRIN2B(24410), GRIN2C(24411), GRIN2D(24412), GRIN3A(191573), GRIN3B(170796), GRINA(266668)

description

Drug Control: Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet

description

-

description

-

description

-

solubility

H2O: >10 mg/mL

solubility

-

solubility

-

solubility

-

drug control

Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet

drug control

-

drug control

-

drug control

-


Still not finding the right product?

Explore all of our products under Dextrorphan tartrate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters