Skip to Content
Merck

Skip To

CS0008

Enzymatic Protein Biotinylation Kit

for biotinylation of up to 20 mg target protein

Synonym(s):

Biotin labeling kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.84
UNSPSC Code:
12161503
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

shipped in

dry ice

storage temp.

−70°C

General description

Biotinylated proteins are used in a variety of applications, such as binding assays, biopanning, and protein purification. Although chemical biotinylation methods are common, they present several challenges, including partial labeling and high batch to-batch variation, because of the random nature of these methods. In addition, the lack of control of the biotinylation site might affect the biotinylated protein′s activity. The Enzymatic Protein Biotinylation Kit allows direct and specific biotinylation using a specific biotin ligation enzyme. The kit contains all the components required for in vitro biotinylation of up to 20 mg recombinant target protein. Biotinylation is mediated by the enzyme BirA biotin ligase, which uniformly ligates biotin to lysine residues in the context of several tags, such as AviTag and Biotag. This kit was tested successfully under a wide range of conditions, including:
  • Temperatures ranging from 4 °C to 30 °C
  • pH from 6.0 to 9.0
  • NaCl concentrations from 50 to 500 mM.
The kit includes streptavidin, which can be used to assess the degree of biotinylation. The kit also includes a BLPR-tagged protein as a positive control.
Supportive calculator (Click here to download a calculator excel file): Will calculate the reagents needed according to your experimental needs.

Legal Information

AviTag is a trademark of Avidity, LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EDEGLYBTAGBK101
shipped in

dry ice

shipped in

wet ice

shipped in

-

shipped in

-

storage temp.

−70°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–5 of 5 Questions  
  1. Is this product Animal Component Free?

    1 answer
    1. This product is typically not processed using animal component materials. This BirA Ligase is recombinant from E. coli. However, the sources of these materials may change from lot to lot and is reported on the Certificate of Origin. To check the source of a current lot, we kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Documentation" under the Products Section with all the required information so that a member of our team can reach out to you to assist further. Thank you.

      Helpful?

  2. Does the BirA ligase contain a His tag?

    1 answer
    1. The product CS0008A BirA ligase has no tags.

      Helpful?

  3. Is the biotinylation kit compatible with 500 mM imidazole in the sample buffer?

    1 answer
    1. The Enzymatic Protein Biotinylation Kit has not been tested with buffers containing imidazole, including 500 mM imidazole. The kit has been evaluated under a broad range of conditions, including pH 6.0–9.0 and NaCl concentrations from 50 mM to 500 mM. Compatible buffers include MES, Tris, Bicine, and PBS. Therefore, compatibility or performance in the presence of 500 mM imidazole cannot be guaranteed. It is advisable to test the kit with the buffer, possibly at different imidazole concentrations, although optimal results are not assured.

      Helpful?

  4. The gel for product #CS0008 suggests a similar activity of the kit at different pH values (6 - 9). However, in my hands, the reaction proceeds significantly faster at pH 8.3 compared to pH 6/7. Which conditions were used for the shown labeling reactions?

    1 answer
    1. The labeling reactions for this kit were performed at pH 7.

      Helpful?

  5. Is the BirA protein tagged in any way (ideally FLAG) in order to remove it from the reaction mixture?

    1 answer
    1. The BirA ligase is not tagged. There is no suggested protocol for the removal of excess ligase. However, in the presence of ATP and biotin the reaction will run to completion. Any residual ligase will remain inert. The BirA ligase has a molecular weight of 33.5 kDa. Note that excess biotin is removed by dialysis. Please see the link below to review the kit Technical Bulletin:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/700/170/cs0008bul-ms.pdf

      Helpful?

Reviews

No rating value

Active Filters