CLS6796610D
Corning® stirrer with digital display
AC/DC input 230 V AC (Supplied with both 3-pin UK plug and 3-pin Euro plug), plate L × W 10 in. × 10 in.
Synonym(s):
magnetic stirrer, magnetic stirrers, stirrers
All Photos(1)
About This Item
packaging
case of 1
manufacturer/tradename
Corning 6796-610D
availability
not available in USA and Canada
parameter
60-1150 rpm speed
plate L × W
10 in. × 10 in.
General description
- Digital LED stir speed display indicates when set speed has been reached
- Microprocessor maintains consistent and repeatable stir speed settings
- Extremely durable and heat resistant Pyroceram glass-ceramic top
- Small footprint and low profile
- Meets UL and cUL standards
Other Notes
individual units are not routinely calibrated, rpm speeds are approximate only
Legal Information
Corning is a registered trademark of Corning, Inc.