Skip to Content
Merck
All Photos(1)

Key Documents

CLS432081

Accessory for Corning® CoolBox

cooling core for Corning® CoolBox XT workstations, for 0.5 to 4 C

Synonym(s):

CoolBox Accessory, CoolBox Kit, Corning CoolBox

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123304
NACRES:
NC.01

feature

cooling core for Corning® CoolBox XT workstations
for 0.5 to 4 C

packaging

pkg of 1

manufacturer/tradename

Corning 432081

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

accessory for coolbox

Legal Information

CoolBox is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service