Skip to Content
Merck
All Photos(1)

Key Documents

CLS42592

Corning® 1 x 8 Stripwell 96 well plates

high binding surface, flat bottom clear (4 cavity), non-sterile

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
flat bottom clear (4 cavity)
flat bottom clear polystyrene (96-well)
size:
96 wells

material

flat bottom clear (4 cavity)
flat bottom clear polystyrene (96-well)

sterility

non-sterile

feature

lid: no

packaging

case of 100
plate of 12 strips

manufacturer/tradename

Corning 42592

size

96 wells

well working volume

75-200 μL , for low volume plates volume is 50-125 μL

suitability

suitable for (cell cuture)

Looking for similar products? Visit Product Comparison Guide

General description

The Costar 1 × 8 strips are pre-assembled in an "egg crate" style 96 well strip holder. Strips fit in holder only one way, eliminating mis-orientation. If covers are needed, the notched cover fits the fully assembled strip holder.

Application

Corning 1 x 8 Stripwell plates have been used extensively in various ELISA and competitive biopanning experiments wherein these plates have been coated with specific antigens to be able to detect and differentiate distinct types of proteins and antibodies.

Features and Benefits

  • Corning 1 x 8 Stripwell plates are made of clear polystyrene.
  • These plates are flat-bottomed and come without a lid.
  • The surface of these plates is well suited for enzyme immunoassay/radioimmunoassay (RIA) tests.
  • The Corning 1 x 8 Stripwell plates are gamma irradiated at a specific dose which has been validated to render the surface effective for strong binding of proteins.

Legal Information

Corning is a registered trademark of Corning, Inc.
Stripwell is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Competitive Panning Method Reveals an Anti-SARS-CoV-2 Nanobody Specific for an RBD-ACE2 Binding Site
Siqi He, et al.
Vaccine (2023)
Development of an ELISA Method to Differentiate Animals Infected with Wild-Type African Swine Fever Viruses and Attenuated HLJ/18-7GD Vaccine Candidate
Lulu Wang, et al.
Viruses (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service