CLS3985BC
Corning® 384 well microplate
Tissue Culture (TC)-treated surface, clear polystyrene, flat bottom, non-sterile, with generic barcode, lid, case of 50
Synonym(s):
384 multiwell plates
About This Item
clear polystyrene
flat bottom
Recommended Products
material
clear bottom
clear polystyrene
flat bottom
sterility
non-sterile
feature
lid
skirt
with generic barcode
packaging
case of 50
manufacturer/tradename
Corning 3985BC
maximum volume
112 μL
size
384 wells
wells
384 wells
working volume
80 μL
color
black
suitability
suitable for (cell culture)
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service