Skip to Content
Merck
All Photos(1)

Key Documents

CLS3736

Corning ®Costar® Not Treated Multiple Well Plates

size 6 wells, clear, polystyrene, flat bottom, case of 100 (20 Bulk Packs of 5), sterile, lid

Synonym(s):

cell culture plate, multi well plates, multiwell plates, not treated, tissue culture plates, well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.13
material:
clear
clear bottom
flat bottom
polystyrene
size:
6 wells
sterility:
sterile; γ-irradiated by SER-TAIN process
sterile
binding type:
non-treated surface
feature:
lid
skirt
Pricing and availability is not currently available.

material

clear
clear bottom
flat bottom
polystyrene

sterility

sterile; γ-irradiated by SER-TAIN process
sterile

feature

lid
skirt

packaging

case of 100 (20 Bulk Packs of 5)
pkg of bulk pack

manufacturer/tradename

Corning 3736

size

6 wells

well volume

16.8 mL

well working volume

1.9-2.9 mL

color

clear

suitability

suitable for (cell culture)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Flat bottoms
  • Nonreversible lids with condensation rings to reduce contamination
  • Individual alphanumerical codes for well identification
  • Uniform footprint for ease in stacking
  • Certified nonpyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.
SER-TAIN is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service