CLS357525
Corning® Falcon® Serological Pipettes
capacity 25 mL, individually wrapped, space saver, sterile
Synonym(s):
pipet
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.02
Recommended Products
material
polystyrene
sterility
sterile
feature
individually wrapped space saver
subdivision
manufacturer/tradename
Corning 357525
capacity
25 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Liquid Handling
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service