Skip to Content
Merck
All Photos(1)

Key Documents

CLS353296

Corning® Falcon® Microplate

wells, 96, Tissue Culture (TC)-treated surface, flat bottom white wells, sterile

Synonym(s):

Microplate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

flat bottom white wells

sterility

sterile

feature

lid
wells 96

packaging

case of 50 ea (10 packs of 5)

manufacturer/tradename

Corning 353296

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Falcon® 96 Well White Flat Bottom TC-Treated Microtest Assay Plate, with Lid, Sterile, 5/Pack, 50/Case

Application

Assay

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service