Skip to Content
Merck

CLS352002

Corning® Falcon® Polypropylene Round Bottom Test Tubes

capacity 5 mL, cap: no, non-sterile, case of 1000

Synonym(s):

culture tube

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.13
UNSPSC Code:
41121700
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

round polypropylene

sterility

non-sterile

feature

cap: no

packaging

case of 1000

manufacturer/tradename

Corning 352002

capacity

5 mL

Application

Liquid Handling

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library