Skip to Content
Merck
All Photos(1)

Key Documents

CLS3472

Corning® Transwell® 24 well plates

3.0 μm pore size, Tissue Culture (TC)-treated surface round , clear polystyrene plate, polyester membrane (Polyethylene Terephthalate (PET), flat bottom wells clear sterile, case of 12, pack of 6, 48 ea

Synonym(s):

cell culture inserts, plate inserts, tissue culture dishes, tissue culture plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.13

Product Name

Corning® Transwell® polyester membrane cell culture inserts, 6.5 mm Transwell with 3.0 μm pore polyester membrane insert, TC-treated, sterile, 48/cs

material

clear polystyrene plate
flat bottom wells clear
polyester membrane (Polyethylene Terephthalate (PET)
round

sterility

sterile

feature

lid
skirt

packaging

case of 12
pack of 6

manufacturer/tradename

Corning 3472

insert working volume

0.1 mL

membrane diam.

6.5 mm

membrane thickness

10 μm

size

24 wells

surface area

0.33 cm2 , cell growth area

well working volume

0.6 mL

color

clear

pore size

3.0 μm

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • 10μm thick transparent polyester membrane
  • Treated for optimal cell attachment
  • Packaged 12 inserts in a 24 well plate, 4 plates per case
  • Excellent visibility under phase contrast microscopy
  • Sterilized by gamma radiation
Transwell cell culture inserts are convenient, easy-to-use permeable support devices for the study of both anchorage-dependent and anchorage-independent cell lines They feature a 10 mm, thin, microscopically transparent polyester membrane that is tissue culture treated for optimal cell attachment and growth.

Provides excellent cell visibility under phase contrast microscopy and allows assessment of cell viability and monolayer formation.

Legal Information

Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc P Hübner et al.
PLoS neglected tropical diseases, 14(7), e0008427-e0008427 (2020-07-07)
A major impediment to eliminate lymphatic filariasis and onchocerciasis is the lack of effective short-course macrofilaricidal drugs or regimens that are proven to be safe for both infections. In this study we tested oxfendazole, an anthelmintic shown to be well
Janet L Markman et al.
Nature communications, 11(1), 1613-1613 (2020-04-03)
In men, the incidence of melanoma rises rapidly after age 50, and nearly two thirds of melanoma deaths are male. The immune system is known to play a key role in controlling the growth and spread of malignancies, but whether
Youssef Abdalla et al.
Journal of nanobiotechnology, 19(1), 60-60 (2021-02-28)
Approximately 80% of brain tumours are gliomas. Despite treatment, patient mortality remains high due to local metastasis and relapse. It has been shown that transferrin-functionalised porous silicon nanoparticles (Tf@pSiNPs) can inhibit the migration of U87 glioma cells. However, the underlying
Christoph Malkmus et al.
PLoS neglected tropical diseases, 14(11), e0008503-e0008503 (2020-11-06)
Onchocerciasis also known as river blindness is a neglected tropical disease and the world's second-leading infectious cause of blindness in humans; it is caused by Onchocerca volvulus. Current treatment with ivermectin targets microfilariae and transmission and does not kill the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service