Skip to Content
Merck

CLS3391

Corning® HTS Transwell® 96 well permeable supports

pore size 0.4 μm, polycarbonate membrane

Synonym(s):

corning transwell, transwell corning

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.75
UNSPSC Code:
41121800
Material:
flat bottom wells clear , polycarbonate membrane, polystyrene , round wells
Size:
0.235 mL , 96 wells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat bottom wells clear , polycarbonate membrane, polystyrene , round wells

sterility

sterile

feature

lid, skirt, plate format: 96 well

packaging

case of 5

manufacturer/tradename

Corning 3391

membrane thickness

10.0 μm

size

0.235 mL , 96 wells

color

clear plate

pore size

0.4 μm pore size

suitability

suitable for (cell culture applications; optimal for cell attachment)

binding type

Tissue Culture (TC)-treated surface

General description

  • Product includes Transwell® unit (1 integral tray with 96 wells), reservoir plate (for cell growth steps), receiver plate (96 wells for growth or assay steps) and 2 sterile lids; 5 sets per case
  • 0.4 μm pore size PC (polycarbonate) membrane
  • Membrane is treated for optimal cell attachment
  • Cell growth area is 0.143 cm² which is 20-50% greater than other brands
  • Large apical and basolateral access ports
  • Automation optimized design with multi-channel feeder ports, improved gripping surface and bar coding

Legal Information

Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library