Skip to Content
Merck

LDS 73/LW1

96050401, Reactivity: Anti-Glutathione S-Transferase subunit YC2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
Biological source:
Mouse x Mouse Hybridoma
Isotype:
IgG2a
Technique(s):
cell culture | mammalian: suitable
Myeloma:
NS0
Reactivity:
Anti-Glutathione S-Transferase subunit YC2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Mouse x Mouse Hybridoma

ImmunoDon

MF1 x BALB/c F mouse spleen cells

Myeloma

NS0

Reactivity

Anti-Glutathione S-Transferase subunit YC2

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Immunogen

Rat glutathione S-transferase subunit YC2

Application

Immunohistochemistry, Western blotting, ELISA assays

Biochem/physiol Actions

Mouse NS0 x Mouse (MF1xBALB/c F) spleen
The hybridoma cell line LDS 73/LW1 was produced by fusing NS0 myeloma cells with spleen cells from an MF1 x BALB/c mouse immunised with rat glutathione S-transferase subunit YC2. The cell line, also known as LW1, secretes antibodies that distinguish between subunit YC2 and YC1.

Preparation Note

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C
RPMI 1640 or DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). Use of this product by commercial organizations may require a license agreement. Commercial customers will be contacted by [email protected] to provide further information after the order request is submitted. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to [email protected].

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library