Skip to Content
Merck
All Photos(1)

Documents

MOR/CPR

96042333, human lung, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MOR/CPR, 96042333

biological source

human lung

growth mode

Aggregates in suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lung adenocarcinoma, drug-resistant

Cell Line Description

MOR/CPR has been developed by growing the parent line, MOR, in increasing concentrations of cisplatin. The cells are cross-resistant to melphalan but show little or no cross-resistance to other platinum compounds. Cisplatin accumulation has been found to be reduced compared to the parent line.

Application

Analysis of acquired drug resistance

Culture Medium

RPMI 1640 + 2mM Glutamine + 1μg/ml cisplatin + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split subconfluent cultures (70-80%) 1:2 to 1:4 seeding at 2-4 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. It is recommended to culture the cells without drug after resuscitation until the first passage. These cells produce a

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service