Skip to Content
Merck
All Photos(1)

Key Documents

EMT6/MTXR

96042322, mouse unknown/unspecified, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

EMT6/MTXR, 96042322

biological source

mouse unknown/unspecified

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse mammary, drug-resistant

Cell Line Description

The drug-resistant cell line EMT6/MTXR was derived from the mouse mammary tumour cell line EMT6/P (Sigma Catalogue number. 96042344) by growth in increasing concentrations of methotrexate. EMT6/MTXR did not hyperexpress P-glycoprotein and did not show cross-resistance to other drugs of the multi-drug resistance (MDR) phenotype. It is recommended to culture the cells without drug on resuscitation until first passage or growth has been fully established.

Application

Drug-resistance studies

Culture Medium

MEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 1μg/ml methotrexate + 15% Foetal Bovine Serum (FBS).. Resuscitate in drug-free medium; add drug once growth is fully established

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 using 0.25% trypsin or trypsin/EDTA;5% CO2; 37ºC

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service