Skip to Content
Merck

NCI-H510A

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
Biological source:
human lung (adrenal metastasis)
Relevant disease(s):
metastasis
Growth mode:
Adherent/aggregates in suspension
Karyotype:
Hypotriploid; modal number = 54; range = 46 to 57. Twenty to 25 marker chromosomes common to all cells. These include t(13q21q), der(1)t(1;21)(p36.1;q11), der(7)t(7;7)(p22;q22), 11p+, 12p+, and 15p+.
Morphology:
Not specified
Manufacturer/tradename:
ECACC
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human lung (adrenal metastasis)

manufacturer/tradename

ECACC

packaging

tube of 5 μg 96020944-DNA-5UG, pkg of vial of cells 96020944-1VL

growth mode

Adherent/aggregates in suspension

karyotype

Hypotriploid; modal number = 54; range = 46 to 57. Twenty to 25 marker chromosomes common to all cells. These include t(13q21q), der(1)t(1;21)(p36.1;q11), der(7)t(7;7)(p22;q22), 11p+, 12p+, and 15p+.

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Biochem/physiol Actions

Human lung carcinoma, adrenal gland metastasis
The NCI-H510A cell line was derived by D. Carney, A.F. Gazdar amd associates in 1982 from an adrenal metastasis in an adult, 56-year-old, male Caucasian patient. The cells produce easily detectable p53 mRNA at levels comparable to those in normal lung tissue. The line does not exhibit any gross structural DNA abnormalities. The cells express elevated levels of four biochemical markers of small cell lung carcinoma (SCLC) i.e. neuron specific enolase, the brain isoenzyme creatine kinase, L-DOPA decarboxylase and bombesin-like immunoreactivity. This cell line is tumourigenic in nude mice (the cells form transplantable tumours with typical small cell carcinoma histology).

Preparation Note

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Split sub-confluent cultures (70-80%) 1:3 to 1:8 i.e. seeding at 2-4x10,000 cells/cm2; 5% CO2; 37°C. This cell line grows as a mixture of cells in suspension and adherent cells. Subcultures can be prepared by scraping the adherent cells into the medium,

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library