OV7
96020764, human ovary, Epithelial
Synonym(s):
OV-7 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
OV7, 96020764
biological source
human ovary
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
VEGF
receptors
VEGF receptors flt and KDR
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human ovarian cancer, stage III
Cell Line Description
The OV7 cell line was established from solid ovarian tumour tissue taken from a 78-year-old female patient. The tumour was characterised as stage III, mixed tumour type and poorly differentiated. Immunocytochemical staining of the cells was reported to show strong staining for cytokeratin (CK), no staining for carcinoembryonic antigen (CEA) and strong staining for human milk fat globulin (HMFG-2). Detection of mRNA expression of vascular endothelial growth factor (VEGF) and its receptors flt and KDR has been reported. The patient had received treatment with chlorambucil.
Culture Medium
DMEM:HAMS F12 (1:1) + 2mM Glutamine + 5% Foetal Bovine Serum (FBS) + 0.5 ug/ml hydrocortisone + 10ug/ml insulin
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service