Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Specific activity:
≥100 units/mg protein
Pricing and availability is not currently available.
Application
Creatine kinase MM fraction from human heart has been used in a study to investigate the importance of intraoperative and postoperative cardiac medical therapy in emergency coronary artery bypass grafting for acute myocardial infarction. Creatine kinase MM fraction from human heart has also been used in a study to investigate the circadian dependence of infarct size and left ventricular function after ST slevation myocardial infarction.
Biochem/physiol Actions
May be used as a control or calibrator in monitoring myocardial injury.
One unit will transfer 1.0 μmole of phosphate from creatine phosphate to ADP per minute at 37 °C (measured at 340 nm as one equimolar amount of NADH produced by coupled reaction).
Physical form
Lyophilized powder containing Tris-HCl, EDTA and N-acetyl cysteine.
Other Notes
One unit will transfer one micromole of phosphate from creatine phosphate to ADP per minute at 37 deg C.
Disclaimer
All human source materials have tested negative for HIV1, HIV2, HCV antibodies and HbsAg. No test guarantees a product to be non-infectious. Therefore, all material derived from human fluids or tissues should be considered as potentially infectious.
1 of 1
This Item | |||
|---|---|---|---|
| Gene Information human ... CKM(1158) | Gene Information human ... CKM(1158) | Gene Information human ... CKM(1158) | Gene Information human ... CKM(1158) |
| specific activity ≥100 units/mg protein | specific activity - | specific activity - | specific activity - |
| form lyophilized powder | form buffered aqueous solution | form buffered aqueous solution | form buffered aqueous solution |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| Quality Level 200 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the Department of Transportation shipping information for this product?
1 answer-
Helpful?
-
-
What does the MM stand for in Product C9858, Creatine Kinase MM Fraction from human heart?
1 answer-
Creatine Kinase is a dimer that exists as three isozymes: MM (muscle), MB (hybrid) and BB (brain).
Helpful?
-



