Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C26H52NO6P
Molecular Weight:
505.67
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352211
MDL number:
Assay:
≥93% (HPLC)
Form:
film, powder or solid
Quality Segment
assay
≥93% (HPLC)
form
film, powder or solid
color
white to off-white
solubility
DMSO: 15 mg/mL, ethanol: 20 mg/mL, clear to hazy (Colorless to faint yellow)
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](COP(O)(O)=O)NC(=O)CCCCCCC
InChI
1S/C26H52NO6P/c1-3-5-7-9-10-11-12-13-14-15-16-18-19-21-25(28)24(23-33-34(30,31)32)27-26(29)22-20-17-8-6-4-2/h19,21,24-25,28H,3-18,20,22-23H2,1-2H3,(H,27,29)(H2,30,31,32)/b21-19+/t24-,25+/m0/s1
InChI key
VSSNYUXSRXINIP-WRBRXSDHSA-N
Biochem/physiol Actions
Cell permeable stimulator of DNA synthesis in cultured fibroblasts; antagonizes mitogenic activity.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C8355-1MG | 04061826674307 |