Skip to Content
Merck

C7927

Cecropin P1 Porcine

≥95% (HPLC), powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C147H253N45O43
CAS Number:
Molecular Weight:
3338.86
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

antibiotic activity spectrum

fungi

mode of action

cell membrane | interferes

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)CNC(=O)C(CCC(O)=O)NC(=O)C(CO)NC(=O)C(NC(=O)C(CCCNC(N)=N)NC(=O)C(CCCCN)NC(=O)C(CCCCN)NC(=O)C(C)NC(=O)C(CO)NC(=O)C(CC(N)=O)NC(=O)C(CCC(O)=O)NC(=O)C(CC(C)C)NC(=O)C(CCCCN)NC(=O)C(CCCCN)NC(=O)C(C)NC(=O)C(NC(=O)C(CCCCN)NC(=O)C(CO)NC(=O)C(CC(C)C)NC(=O)C(Cc1c[nH]c2ccccc12)NC(=O)C(N)CO)C(C)O)C(C)CC)C(=O)NC(C)C(=O)NC(C(C)CC)C(=O)NC(C)C(=O)NC(C(C)CC)C(=O)NC(CCC(N)=O)C(=O)NCC(=O)NCC(=O)N3CCCC3C(=O)NC(CCCNC(N)=N)C(O)=O

InChI

1S/C147H253N45O43/c1-18-75(9)113(140(229)167-81(15)120(209)188-114(76(10)19-2)141(230)168-82(16)121(210)189-115(77(11)20-3)142(231)178-94(47-50-106(154)198)123(212)164-66-108(200)163-68-110(202)192-60-36-46-105(192)139(228)179-97(145(234)235)45-35-59-161-147(158)159)187-109(201)67-165-124(213)95(48-51-111(203)204)176-138(227)104(72-196)186-143(232)116(78(12)21-4)190-130(219)93(44-34-58-160-146(156)157)174-127(216)90(41-26-31-55-150)172-125(214)88(39-24-29-53-148)170-118(207)79(13)166-136(225)102(70-194)184-135(224)101(64-107(155)199)183-129(218)96(49-52-112(205)206)177-132(221)98(61-73(5)6)181-128(217)91(42-27-32-56-151)173-126(215)89(40-25-30-54-149)171-119(208)80(14)169-144(233)117(83(17)197)191-131(220)92(43-28-33-57-152)175-137(226)103(71-195)185-133(222)99(62-74(7)8)182-134(223)100(180-122(211)86(153)69-193)63-84-65-162-87-38-23-22-37-85(84)87/h22-23,37-38,65,73-83,86,88-105,113-117,162,193-197H,18-21,24-36,39-64,66-72,148-153H2,1-17H3,(H2,154,198)(H2,155,199)(H,163,200)(H,164,212)(H,165,213)(H,166,225)(H,167,229)(H,168,230)(H,169,233)(H,170,207)(H,171,208)(H,172,214)(H,173,215)(H,174,216)(H,175,226)(H,176,227)(H,177,221)(H,178,231)(H,179,228)(H,180,211)(H,181,217)(H,182,223)(H,183,218)(H,184,224)(H,185,222)(H,186,232)(H,187,201)(H,188,209)(H,189,210)(H,190,219)(H,191,220)(H,203,204)(H,205,206)(H,234,235)(H4,156,157,160)(H4,158,159,161)

InChI key

PRIVBYDFWSFUFP-UHFFFAOYSA-N

General description

Cecropin P1 is found in Ascaris suum, which is a parasite that lives in porcine intestine.

Application

Cecropin P1 Porcine has been used to study its antimicrobial activity against Escherichia coli.

Biochem/physiol Actions

Antibacterial peptide originally identified in moths (Hyalophora cecropia) and later in pig intestine.
Cecropin P1 can inhibit the growth of enterotoxigenic Escherichia coli in porcine. It also can inhibit the growth of human fungal pathogen Candida albicans. Cecropin P1 also blocks the release of viral particles and impairs virus-induced apoptosis. Cecropins are cationic anti-microbial peptides (AMPs). It can inactivate microorganisms by forming an α-helical chain which punctures the cell membrane. Cecropins may be used in food preservation as it is non-cytotoxic to mammalian cells.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library