Skip to Content
Merck
All Photos(2)

Documents

C7082

Sigma-Aldrich

Monoclonal Anti-β-Catenin antibody produced in mouse

clone 6F9, ascites fluid

Synonym(s):

Anti-CTNNB, Anti-EVR7, Anti-MRD19, Anti-NEDSDV, Anti-armadillo

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

6F9, monoclonal

mol wt

antigen 94 kDa

contains

15 mM sodium azide

species reactivity

human, chicken, canine, bovine

technique(s)

indirect immunofluorescence: 1:1,000 using cultured MDBK cells
microarray: suitable
western blot: 1:1,000 using cultured MDBK cells extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTNNB1(1499)

General description

Monoclonal Anti- β -Catenin (mouse IgG1 isotype) is derived from the 6F9 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. CTNNB1 (catenin β 1) gene is mapped to human chromosome 3p21. The protein encoded is also called as cadherin-associated protein.The encoded protein β-catenin is localized to the nucleus. Monoclonal antibody reacting specifically with β -catenin is an essential tool in defining the interactions and distributions of β -catenin and its relationships with other catenins and cadherins in various cells and tissues.

Specificity

The antibody does not cross-react with plakoglobin (γ-catenin).

Immunogen

recombinant chicken β-catenin.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Monoclonal Anti-β-Catenin antibody produced in mouse has been used in:
  • immunohistochemistry
  • immunofluorescence
  • immunoblotting

Biochem/physiol Actions

In Wnt/β-catenin signal pathway, β-Catenin participates as a signal transduction molecule. The pathway is associated with development, tissue homeostasis and oncogenic transcription. Upregulation of the gene is associated with glioma cell proliferation. The encoded protein has dual function that regulates the coordination of cell-cell adhesion by linking cytoskeleton and cellular junctions and gene transcription. Wnt/β-catenin signal pathway is known to induce progression of colon cancer progression. It is useful for differential diagnosis of Osteoblastoma (OB) and osteosarcoma (OS). β-catenin signaling pathway may contribute to cholangiocarcinoma (CCA) cell proliferation. Monoclonal antibody reacting specifically with β -catenin is an essential tool in defining the interactions and distributions of β -catenin and its relationships with other catenins and cadherins in various cells and tissues.

Physical form

Supplied as ascites fluid with 15 mM sodium azide as a preservative.

Storage and Stability

For extended storage freeze in working aliquots. For continuous use, store at 2-8 °C. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suppression of canonical Wnt/?-catenin signaling by nuclear plakoglobin recapitulates phenotype of arrhythmogenic right ventricular cardiomyopathy
Gracia Gras E, et al.
The Journal of Clinical Investigation (2006)
Jenny Schneider et al.
Development (Cambridge, England), 142(15), 2686-2695 (2015-06-28)
The nephron is the fundamental structural and functional unit of the kidney. Each mature nephron is patterned along a proximal-distal axis, with blood filtered at the proximal end and urine emerging from the distal end. In order to filter the
SOX7 Suppresses Wnt Signaling by Disrupting ?-Catenin/BCL9 Interaction.
Fan R, et al.
Dna and Cell Biology, 37(2), 126-132 (2018)
The protective effect of PPAR? in sepsis-induced acute lung injury via inhibiting PTEN/?-catenin pathway
Liu L, et al.
Bioscience Reports, 40(5) (2020)
Loss of FOXN3 in colon cancer activates beta-catenin/TCF signaling and promotes the growth and migration of cancer cells
Yuedi D, et al.
Oncotarget, 8(6), 9783-9793 (2017)

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service