C7048
Monoclonal Anti-CD3 antibody produced in mouse
clone UCHT-1, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Monoclonal Anti-CD3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
UCHT-1, monoclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
flow cytometry: 5 μL using 1 × 106 cells
isotype
IgG1
UniProt accession no.
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... CD3E(916)
Looking for similar products? Visit Product Comparison Guide
Specificity
Recognizes the CD3 complex. The epitope recognized by clone no. UCHT-1 is expressed on the ε-chain of the CD3 antigen/T cell receptor complex. 3rd Workshop: code no. 126, 208 and 471
4th Workshop: paper no. T3.2
4th Workshop: paper no. T3.2
Immunogen
human thymocytes followed by Sezary T cells.
Application
Monoclonal Anti-CD3 antibody produced in mouse is suitable for flow cytometry at a dilution of 5μL using 1 × 106 cells.
Biochem/physiol Actions
The CD3e cytoplasmic domain of the T cell receptor binds to the inner leaflet of the plasma membrane (PM) and is accessible during early stages of T cell activation. It is specific for T lymphocytes, found in Purkinje cells of the cerebellum and gastric parietal cells of humans and other species. The antibodies commonly used to recognize CD3 are directed against the ε-subunit of the T cell receptor. Cytoplasmic cyCD3 expression can only be achieved during the early phase of NK-cell differentiation.
Target description
The CD3 complex (16/20/29/22/25-28 kDa) consists of 5 polypeptide chains associated with each other and with the T cell receptor.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bernd Schlereth et al.
Cancer research, 65(7), 2882-2889 (2005-04-05)
Bispecific T-cell engager (BiTE) are a class of bispecific single-chain antibodies that can very effectively redirect cytotoxic T cells for killing of tumor target cells. Here, we have assessed the in vivo efficacy of one representative, called bscEp-CAMxCD3, with specificity
Francesco Dieli et al.
The Journal of infectious diseases, 186(12), 1835-1839 (2002-11-26)
Vgamma9/Vdelta2 T cells can contribute to protective immune response against Mycobacterium tuberculosis, although the extent to which and mechanisms by which they could actually protect against human tuberculosis remain unclear. We have previously reported that Vgamma9/Vdelta2 T cells from tuberculin
F Dieli et al.
The Journal of infectious diseases, 181(1), 294-301 (1999-12-23)
The alphabeta and gammadelta T cell responses were analyzed in the peripheral blood of children affected by active tuberculosis (TB) and in healthy children who tested positive (PPD+) or negative (PPD-) for purified protein derivative. PPD+ healthy and diseased children
Graeme O'Boyle et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(12), 4598-4603 (2012-03-07)
The recruitment of T lymphocytes during diseases such as rheumatoid arthritis is regulated by stimulation of the chemokine receptors expressed by these cells. This study was designed to assess the potential of a CXCR3-specific small-molecule agonist to inhibit the migration
Joseph Alroy et al.
Pathology, research and practice, 201(12), 803-808 (2005-11-26)
CD3 antigen, formerly thought to be specific for T lymphocytes, has been identified in Purkinje cells of the cerebellum and gastric parietal cells in several species, including humans. The antibodies commonly used to recognize CD3 are directed against the epsilon-subunit
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service