Skip to Content
Merck
All Photos(1)

Key Documents

C6706

Sigma-Aldrich

Z-Leu-Leu-Norvalinal

≥90% (HPLC), powder

Synonym(s):

MG115, Z-LLnV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H39N3O5
CAS Number:
Molecular Weight:
461.59
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Assay

≥90% (HPLC)

form

powder

color

white

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

CCC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(=O)OCc1ccccc1)C=O

InChI

1S/C25H39N3O5/c1-6-10-20(15-29)26-23(30)21(13-17(2)3)27-24(31)22(14-18(4)5)28-25(32)33-16-19-11-8-7-9-12-19/h7-9,11-12,15,17-18,20-22H,6,10,13-14,16H2,1-5H3,(H,26,30)(H,27,31)(H,28,32)/t20-,21-,22-/m0/s1

InChI key

QEJRGURBLQWEOU-FKBYEOEOSA-N

Amino Acid Sequence

Z-Leu-Leu-Nva

Biochem/physiol Actions

Potent inhibitor of the 20S proteasome (Ki = 21 nM) and 26S proteasome (Ki = 35 nM). Cells treated with 54 μM N-CBZ-Leu-Leu-Norvalinal showed complete block of the G1/S and metaphase transitions and delayed passage through S-phase. Induces apoptosis in Rat-1 and PC12 cells, perhaps by accumulation of stabilized p53.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Gangwei Ou et al.
PloS one, 4(11), e7806-e7806 (2009-11-13)
Vibrio cholerae is the causal intestinal pathogen of the diarrheal disease cholera. It secretes the protease PrtV, which protects the bacterium from invertebrate predators but reduces the ability of Vibrio-secreted factor(s) to induce interleukin-8 (IL-8) production by human intestinal epithelial
U G Lopes et al.
The Journal of biological chemistry, 272(20), 12893-12896 (1997-05-16)
Proteolysis by the ubiquitin/proteasome pathway controls the intracellular levels of a number of proteins that regulate cell proliferation and cell cycle progression. To determine whether this pathway of protein turnover was also linked to apoptosis, we treated Rat-1 and PC12
Brooke A Jude et al.
Journal of bacteriology, 191(22), 6911-6917 (2009-09-08)
Vibrio cholerae is the etiologic agent of cholera in humans. Intestinal colonization occurs in a stepwise fashion, initiating with attachment to the small intestinal epithelium. This attachment is followed by expression of the toxin-coregulated pilus, microcolony formation, and cholera toxin
Yongjian Qiu et al.
Nature communications, 8(1), 1905-1905 (2017-12-05)
Plant phytochromes are thought to transduce light signals by mediating the degradation of phytochrome-interacting transcription factors (PIFs) through the N-terminal photosensory module, while the C-terminal module, including a histidine kinase-related domain (HKRD), does not participate in signaling. Here we show
Takahiko Ishikawa et al.
PloS one, 4(8), e6734-e6734 (2009-08-25)
The type VI secretion system (T6SS) has emerged as a protein secretion system important to several gram-negative bacterial species. One of the common components of the system is Hcp, initially described as a hemolysin co-regulated protein in a serotype O17

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service