Skip to Content
Merck

Skip To

C6368

β-Cryptoxanthin

≥97% (TLC)

Synonym(s):

Caricaxanthin, Hydroxy-β-carotene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C40H56O
CAS Number:
Molecular Weight:
552.87
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (TLC)

form

powder

color

faint red to very dark red

mp

169-170 °C

solubility

chloroform: 1 mg/mL

storage temp.

−20°C

SMILES string

CC(=C\C=C\C=C(C)\C=C\C=C(C)\C=C\C1=C(C)C[C@@H](O)CC1(C)C)/C=C/C=C(C)/C=C/C2=C(C)CCCC2(C)C

InChI

1S/C40H56O/c1-30(18-13-20-32(3)23-25-37-34(5)22-15-27-39(37,7)8)16-11-12-17-31(2)19-14-21-33(4)24-26-38-35(6)28-36(41)29-40(38,9)10/h11-14,16-21,23-26,36,41H,15,22,27-29H2,1-10H3/b12-11+,18-13+,19-14+,25-23+,26-24+,30-16+,31-17+,32-20+,33-21+/t36-/m1/s1

InChI key

DMASLKHVQRHNES-FKKUPVFPSA-N

General description

β-Cryptoxanthin is an oxygen-containing carotenoid and is a member of the xanthophyll family. It is one of the major sources of vitamin A and is present in fruits such as oranges, tangerines and papayas. In addition, it is also found in corn, peas and some animal products such as egg yolk and butter.

Application

β-Cryptoxanthin has also been used to study its effect on the production of immunoglobulins in Peyer′s patch cells ex- vivo. It has also been used as a standard in high-performance liquid chromatography (HPLC analysis).(5)

Biochem/physiol Actions

Carotenoid pigment with antioxidant functionality. In vivo conversion of β-cryptoxanthin to retinol can occur in humans. Inhibits carcinogenic induced urinary bladder cancer in experimental animal models.
β-Cryptoxanthin exhibits potential-anabolic effect on bone calcification by stimulating osteoblastic bone formation and inhibiting osteoclastic bone resorption in vitro. It acts as an antioxidant and avoids free radical damage to biomolecules such as lipids, proteins and nucleic acids. High dietary intake of β-cryptoxanthin reduces the risk of developing rheumatoid arthritis and lung cancer.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C9750SML098269553
assay

≥97% (TLC)

assay

≥93% (UV)

assay

≥95% (HPLC)

assay

≥90.0% (HPLC)

form

powder

form

powder

form

powder

form

solid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

solubility

chloroform: 1 mg/mL

solubility

hexane: 1.1 mg/mL, DMSO: 30 μg/mL

solubility

DMSO: 1 mg/mL (warmed)

solubility

-

color

faint red to very dark red

color

dark red to brown

color

, pink to very dark purple

color

-

mp

169-170 °C

mp

178-179 °C

mp

-

mp

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters