Skip to Content
Merck
All Photos(1)

Key Documents

C6295

Sigma-Aldrich

Cell Freezing Medium-DMSO Serum free 1x

sterile-filtered, suitable for cell culture

Synonym(s):

cryopreservation medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable
cryopreservation: suitable

impurities

endotoxin, tested

shipped in

dry ice

storage temp.

−20°C

General description

Cell freezing medium is a combination of cryoprotectant DMSO with fetal bovine serum. However serum free medium replaces serum with methylcellulose and poloxamer 188 and other additives like ascorbic acid, α-tocopherol and proline.

Application

Cell Freezing Medium-DMSO Serum free 1x has been used in the freezing of human neural progenitor cells (hNPCs), olfactory ensheathing cells (OECs) and retinal pigment epithelial (RPE) cells.

Other Notes

Complete ready-to-use medium containing Minimum Essential Medium, dimethyl sulfoxide (8.7%) and methyl cellulose. Prepared according to the formulation of Waymouth.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen-Yang Meng et al.
Oncology reports, 43(4), 1169-1186 (2020-04-24)
Osteosarcoma (OS) is the most common primary malignant tumor of the bone affecting children and adolescents. Chemotherapy is now considered as a standard component of OS treatment, not only for children, but also for adults. However, chemoresistance continues to pose
Acute and delayed transplantation of olfactory ensheathing cells promote partial recovery after complete transection of the spinal cord
Lopez-Vales R, et al.
Neurobiology of Disease, 21(1), 57-68 (2006)
Cryopreservation of cells using defined serum-free cryoprotective agents
Volbers JC, et al.
Current Directions in Biomedical Engineering, 2(1), 315-318 (2016)
Simple freezing procedure for storage in serum-free media of cultured and tumor cells of mouse.
Waymouth, C. and Varnum, D.
TCA Manual, 2(1), 311-313 (1976)
Kai Pollard et al.
Scientific data, 7(1), 184-184 (2020-06-21)
Nerve sheath tumors occur as a heterogeneous group of neoplasms in patients with neurofibromatosis type 1 (NF1). The malignant form represents the most common cause of death in people with NF1, and even when benign, these tumors can result in

Articles

Overview of good cell banking practices for cell line cryopreservation purposes.

Overview of good cell banking practices for cell line cryopreservation purposes.

Overview of good cell banking practices for cell line cryopreservation purposes.

Overview of good cell banking practices for cell line cryopreservation purposes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service