Skip to Content
Merck
All Photos(1)

Key Documents

C5867

Sigma-Aldrich

Anti-Ciliated Cell Marker antibody, Mouse monoclonal

clone LhS 28, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LhS 28, monoclonal

form

buffered aqueous solution

mol wt

antigen ~45 kDa

species reactivity

human, hamster

concentration

~2 mg/mL

technique(s)

electron microscopy: suitable
immunocytochemistry: 1-2 μg/mL using BHK a21 cells
immunohistochemistry: suitable
microarray: suitable
western blot: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Ciliated Cell Marker antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the hybridoma LhS 28 produced by the fusion of mouse myeloma cells (Sp2/0 cells) and splenocytes from BALB/c mice.

Specificity

Anti-Ciliated Cell Marker antibody, Mouse monoclonal recognizes an antigen in human and hamster ciliated cells.

Immunogen

BHK a21 cytoskeletal preparation.

Application

Anti-Ciliated Cell Marker antibody, Mouse monoclonal may be used in:
  • immunoelectron microscopy
  • immunoblotting
  • immunohistochemistry
  • immunocytochemistry

May be used in the detection of ciliated cells and for investigating mechanisms of ciliogenesis.

Biochem/physiol Actions

The mouse hybridoma LhS 28 secretes a monoclonal antibody that recognizes the antigen 44-45 kDa protein associated with the ciliary basal bodies. The antigen recognized by LhS 28 is present in the 9+0 microtubule arrays in the basal body structure of the cilium. It serves as an unequivocal marker for in situ detection and in vitro studies of human ciliated epithelial cells. LhS28 is a potential marker for tubal metaplasia identification and may be useful in recognizing low grade cervical glandular intraepithelial neoplasia. It is effective in detecting ciliogenesis and ciliated cells associated with normal and pathological human specimens

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M T Comer et al.
The Histochemical journal, 31(1), 39-43 (1999-07-16)
The process and regulation of ciliogenesis in human epithelia is little understood and many components of the cilium and associated structures have not been characterised. We have identified a monoclonal antibody, LhS28, which recognises a 44,000-45,000 M(r) protein specifically associated
M T Comer et al.
Journal of clinical pathology, 52(5), 355-357 (1999-11-24)
The assessment of neoplastic disease in gynaecological histopathology can be complicated by the high incidence of metaplasia seen in tissues of the female genital tract. There is a need to identify specific tissue markers which can be applied in routine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service