Skip to Content

C4479

9-Cyclopentyladenine monomethanesulfonate

≥98% (HPLC)

Synonym(s):

9-CP-Ade mesylate, 9-Cyclopentyladenine mesylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H13N5 · CH3SO3H
CAS Number:
Molecular Weight:
299.35
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
41106305
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

mp

180-183 °C (lit.)

solubility

H2O: 5 mg/mL, clear

storage temp.

room temp

SMILES string

CS(O)(=O)=O.Nc1ncnc2n(cnc12)C3CCCC3

InChI

1S/C10H13N5.CH4O3S/c11-9-8-10(13-5-12-9)15(6-14-8)7-3-1-2-4-7;1-5(2,3)4/h5-7H,1-4H2,(H2,11,12,13);1H3,(H,2,3,4)

InChI key

AJMDRSJHCYQPQI-UHFFFAOYSA-N

Biochem/physiol Actions

A stable, cell-permeable, non-competitive adenylyl cyclase inhibitor that targets the P-site domain.
A stable, cell-permeable, non-competitive adenylyl cyclase inhibitor that targets the P-site domain. IC50 = 100 μM in detergent-dispersed rat brain preparation.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library