Skip to Content
Merck

C-40000

HEPES Buffered Saline Solution

Hepes BSS (30 mM), 30 ml

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.81
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

aqueous solution

packaging

pkg of 30 ml

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

HEPES Buffered Saline Solution

Application

HEPES Buffered Saline Solution is prepared from raw materials of the highest quality using tissue culture grade water and provided in a ready-to-use format. It is a component in the PromoCell DetachKits designed for the safe and efficient detachment of primary human cells in routine subculturing.

Preparation Note

Click here for more information.
Store HEPES Buffered Saline Solution at -20°C immediately after arrival. If stored properly, the product is stable until the expiry date stated on the label.

Analysis Note

All lots of PromoCell HEPES Buffered Saline Solution are subjected to comprehensive quality control tests. Each lot is routinely tested for absence of cytotoxicity, and physical parameters such as osmolality and pH level. Approved in-house lots are used as a reference. In addition, all lots have been tested for the absence of microbial contaminants (fungi, bacteria).


Still not finding the right product?

Explore all of our products under HEPES Buffered Saline Solution


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library