Skip to Content

C-39620

Follicle Dermal Papilla Cell Growth Medium

SupplementPack containing all media supplements as individual vials, 1 Pack for 500 ml

Synonym(s):

Recommended with C-26500

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.81
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

packaging

pkg of 1 kit

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Follicle Dermal Papilla Cell Growth Medium SupplementPack (basal medium sold separately)

To form a complete medium, it should be combined with the basal medium: C-26500

Application

PromoCell Follicle Dermal Papilla Cell Growth Medium is a low-serum medium that has been developed for the in vitro cultivation of Human Follicle Dermal Papilla Cells. The medium is optimized for primary cultures of fetal, neonatal, and adult tissue, or for the maintenance of follicle dermal papilla cells.Recommended for:Human Follicle Dermal Papilla Cells

Preparation Note

Click here for more information.
Final supplement concentrations after addition to the basal medium:Fetal Calf Serum: 0.04 ml / mlBovine Pituitary Extract: 0.004 ml / mlBasic Fibroblast Growth Factor (recombinant human): 1 ng / ml Insulin: 5 µg / ml
Store the SupplementPack at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Analysis Note

All lots of PromoCell Follicle Dermal Papilla Cell Growth Medium are subjected to comprehensive quality control tests using primary human follicledermal papilla cells. Each lot is checked for growth promoting activity, adherence rate and typical morphology of the tested follicle dermal papilla cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Other Notes

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials


Still not finding the right product?


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library