Skip to Content
Merck
All Photos(2)

Documents

C3930

Sigma-Aldrich

Calmidazolium chloride

solid

Synonym(s):

1-[Bis(4-chlorophenyl)methyl]-3-[2,4-dichloro-β-(2,4-dichlorobenzyloxy)phenethyl]imidazolium chloride, 1-[bis(4-Chlorophenyl)methyl]-3-[2-(2,4-dichlorophenyl)-2-(2,4-dichlorobenzyloxy)ethyl]-1H-imidazolium chloride, R 24571

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H23Cl7N2O
CAS Number:
Molecular Weight:
687.70
Beilstein:
6470244
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

white

storage temp.

2-8°C

SMILES string

[Cl-].Clc1ccc(cc1)C(c2ccc(Cl)cc2)[n+]3ccn(CC(OCc4ccc(Cl)cc4Cl)c5ccc(Cl)cc5Cl)c3

InChI

1S/C31H23Cl6N2O.ClH/c32-23-6-1-20(2-7-23)31(21-3-8-24(33)9-4-21)39-14-13-38(19-39)17-30(27-12-11-26(35)16-29(27)37)40-18-22-5-10-25(34)15-28(22)36;/h1-16,19,30-31H,17-18H2;1H/q+1;/p-1

InChI key

YGEIMSMISRCBFF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitor of calmodulin-regulated enzymes. It is also believed to directly block calcium channels in smooth muscle cells.

Caution

Binds to glass surfaces in aqueous solutions.

Reconstitution

Stock solutions may be prepared in DMSO at 5 mM then diluted to 0.01 mM with buffer. DMSO solutions may be stored for several weeks.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shi-Hong Gu et al.
Journal of insect physiology, 56(1), 93-101 (2009-10-06)
Our previous studies showed that the prothoracicotropic hormone (PTTH) stimulated extracellular signal-regulated kinase (ERK) phosphorylation in prothoracic glands of Bombyx mori both in vitro and in vivo. In the present study, the signaling pathway by which PTTH activates ERK phosphorylation
Mohamad Sabbah et al.
Bioorganic & medicinal chemistry, 19(16), 4868-4875 (2011-07-26)
Thirty N,N'-disubstituted imidazolium salts have been synthesized and evaluated as LuxR antagonists. Substitution on one of the imidazolium nitrogen atoms includes benzhydryl, fluorenyl or cyclopentyl substituent, and alkyl chains of various lengths on the second one. Most of these compounds
Helle Jensen et al.
Molecular immunology, 53(3), 255-264 (2012-09-12)
In this study we demonstrate that histone deacetylase (HDAC)-inhibitor mediated cell surface expression of the structural different NKG2D-ligands MICA/B and ULBP2 is calcium-dependent. Treatment with the calcium chelator EGTA inhibited constitutive as well as HDAC-inhibitor induced MICA/B and ULBP2 cell
Gurpreet Kaur et al.
The Biochemical journal, 430(1), 39-48 (2010-06-10)
The HMG (high-mobility group)-box-containing chromatin-remodelling factor SRY (sex-determining region on the Y chromosome) plays a key role in sex determination. Its role in the nucleus is critically dependent on two NLSs (nuclear localization signals) that flank its HMG domain: the
Débora Mai N Eguti et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 26(4-5), 541-552 (2010-11-11)
We examined the effect of Angiotensin II (Ang II) on the interaction between the Ca(2+)/CaM complex and hNHE1. Considering that calmodulin binds to NHE1 at two sites (A and B), amino acids at both sites were modified and two mutants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service