Skip to Content
Merck

C3868

Anti-Calsequestrin-2 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CASQ2, Anti-fast-twitch, cardiac muscle Calsequestrin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.44
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
8
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~55 kDa

species reactivity

rat, human

species reactivity (predicted by homology)

mouse

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

western blot: 0.5-1.0 μg/mL using HEK-293T cells expressing human calsequestrin-2, western blot: 0.5-1.0 μg/mL using extract of rat cardiac muscle (S1 fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CASQ2(845)
mouse ... Casq2(12373)
rat ... Casq2(29209)

General description

Calsequestrin is a major Ca2+ binding protein in cardiac and skeletal muscle.
Calsequestrin-2 (55 kDa) is found in both sarcoplasmic reticulum′s (SR) terminal cisternae luminal space of both cardiac and in slow skeletal muscle cells. It is made of three domains each with negatively charged thioredoxin-like folds with a β-sheet surrounded by four α-helices.

Immunogen

synthetic peptide corresponding to amino acids 57-74 of rat calsequestrin-2, conjugated to KLH. This sequence is highly conserved in human and rat calsequestrin-2 (single amino acid substitution). This sequence is not found in calsequestrin-1.

Application

Anti-Calsequestrin-2 antibody has been used in immunoblotting, western blotting.
Anti-Calsequestrin-2 is suitable for western blot at a concentration of 0.5-1.0μg/mL using HEK-293T cells expressing human calsequestrin-2 and western blot at a concentration of 0.5-1.0μg/mL using extract of rat cardiac muscle (S1 fraction)

Biochem/physiol Actions

Over expression of cardiac calsequestrin (CASQ2) is associated with depressed cardiovascular function and hypertrophy. Mutations in CASQ2 are thought to be associated with catecholaminergic polymorphic ventricular tachycardia (CPVT). It is a high-capacity, low-affinity Ca2+ binding glycoprotein, which functions as an internal Ca2+ store in the lumen of the sarcoplasmic reticulum (SR).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library