Skip to Content
Merck
All Photos(2)

Key Documents

C3766

Sigma-Aldrich

Cholesterol Esterase from bovine pancreas

lyophilized powder, ≥200 units/g protein

Synonym(s):

Sterol-ester acylhydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

specific activity

≥200 units/g protein

mol wt

84 kDa by SDS-PAGE

composition

protein, ≥20%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Cholesterol Esterase from bovine pancreas is a member of α-β hydrolase superfamily. The core of the protein comprises 11 β-strands, surrounded by 15 α-strands. Catalytic triad is in the centre of the molecule. The C-terminus possesses hydrophobic residues.

Application

Cholesterol Esterase from bovine pancreas has been used:
  • in γ-oryzanol-hydrolyzing activity
  • in cholesterol esterase enzyme activity for quantification of cholesterol and cholesterol esters associated in tear film and lenses samples
  • to test in vitro degradation of poly(fumaroyl bioxirane) maleate (PFM) polymer designed for bone tissue engineering

Cholesterol esterase bound to membrane-associated heparin on brush border membranes aids in the transport of cholesterol and free fatty acid across the membrane. This enzyme is widely used in the determination of serum cholesterol in clinical laboratories. The enzyme from Sigma has been used to evaluate the inhibitory and antioxidant functions of the methanol extract of the Camellia sinensis leaves under in vitro conditions. The enzyme has also been used to digest human serum samples to confirm the presence and position of acyl esters of 7α-hydroxycholesterol.
Enzyme responsible for the hydrolysis of many of the fatty acid esters of cholesterol.
Optimum pH range: 6-8
Activators: Bisphenol A diglycidyl ether, cAMP-dependent protein kinase type II, ethanol, methanol, n-butanol, phosphatidylcholine, phosphatidylethanolamine, sodium taurocholic acid
Inhibitors: Bisphenol A methacrylate, diisopropylfluorophosphate, enolase, Hg2+, sodium fluoride, phosphatidic acid, phosphatidylcholine, phosphatidylserine

Biochem/physiol Actions

Cholesterol esterase (CE) is a reversible enzyme that can hydrolyze or synthesize fatty acid esters of cholesterol and other sterols. Hydrolysis of water insoluble long chain fatty acid esters requires bile salt activation. Hydrolysis of water soluble esters of short chain fatty acids and lysophospholipids does not require activation by bile salts. It also hydrolyzes tri-, di-, and mono-acylglycerols, phospholipids, lysophospholipids, and ceramide. This monomeric glycoprotein may have multiple functions in lipid and lipoprotein metabolism, as well as in atherosclerosis. Its molecular mass is found to be 84 kDa.

Other Notes

Partially purified

Unit Definition

One unit will hydrolyze 1.0 μmole of cholesteryl oleate to cholesterol and oleic acid per minute at pH 7.0 at 37 °C in the presence of taurocholate.

Physical form

This product is partially purified from bovine pancreas and is supplied as a white to light brown lyophilized powder containing ≥20% protein (biuret) and potassium phosphate.

Preparation Note

Cholesterol esterase is soluble in 0.4 M potassium phosphate, pH 7.0 at 1 mg/mL concentration.

Analysis Note

Protein determined by biuret.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Lopez-Candales et al.
Biochemistry, 32(45), 12085-12089 (1993-11-16)
We have recently hypothesized that neutral lipids can, in part, move across biological membranes via a mechanism involving enzymes anchored to membrane proteoglycans such as those found in the brush border of the enterocyte [Bosner, M. S., Gulick, T., Riley
Cholesterol esterase enzyme inhibitory and antioxidant activities of leaves of Camellia sinensis (l) kuntze. using in vitro models
Kumar
International Journal of Pharmaceutical Sciences and Research, 2(11), 2923-2928 (2011)
A functional polymer designed for bone tissue engineering.
You Z, et al.
Acta Biomaterialia, 8(2), 502-510 (2012)
Structure of bovine pancreatic cholesterol esterase at 1.6 ?: novel structural features involved in lipase activation.
Chen JCH, et al.
Biochemistry, 37(15), 5107-5117 (1998)
H Oda et al.
Journal of lipid research, 31(12), 2209-2218 (1990-12-01)
Serum levels of 7 alpha-hydroxycholesterol and activities of hepatic microsomal cholesterol 7 alpha-hydroxylase in surgical patients were analyzed by capillary gas-liquid chromatography-selected ion monitoring technique using a new internal standard, 5 alpha-cholestane-3 beta, 7 beta-diol. We found that concentrations of

Articles

Cholesterol esterification enhances transport efficiency in lipoproteins for increased blood stream transport.

Cholesterol esterification enhances transport efficiency in lipoproteins for increased blood stream transport.

Cholesterol esterification enhances transport efficiency in lipoproteins for increased blood stream transport.

Cholesterol esterification enhances transport efficiency in lipoproteins for increased blood stream transport.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service