Skip to Content
Merck

C3355

Coelenterazine hcp

≥95% (HPLC), solid

Synonym(s):

CLZN-hcp

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H25N3O2
CAS Number:
Molecular Weight:
399.48
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Coelenterazine hcp, solid

assay

≥95% (HPLC)

Quality Segment

form

solid

solubility

methanol: 1 mg/mL, clear, yellow to orange

storage temp.

−20°C

SMILES string

Oc1ccc(cc1)C2=CN3C(=O)C(Cc4ccccc4)=NC3=C(CC5CCCC5)N2

InChI

1S/C25H25N3O2/c29-20-12-10-19(11-13-20)23-16-28-24(21(26-23)14-17-8-4-5-9-17)27-22(25(28)30)15-18-6-2-1-3-7-18/h1-3,6-7,10-13,16-17,26,29H,4-5,8-9,14-15H2

InChI key

YEGCUOQUFAXCMK-UHFFFAOYSA-N

General description

Coelenterazine hcp is a semisynthetic analog of coelenterazine.

Application

Coelenterazine hcp has been used:
  • as a component of coelenterazine reagent in the reporter assay
  • in the purification of Renilla luciferase-labeled Annexin V
  • in evaluating the activity of aequorin in Escherichia coli

Biochem/physiol Actions

Coelenterazine hcp has 190 times higher luminescence intensity than native coelenterazine and a faster response time than native coelenterazine.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library