Skip to Content
Merck

C3160

Complement C5 from human serum

≥90% (SDS-PAGE), >100,000 C5H50 units/mg protein

Synonym(s):

C5 human, Complement C5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.71
MDL number:
Form:
solution
Assay:
≥90% (SDS-PAGE)
Biological source:
human serum
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human serum

Quality Segment

assay

≥90% (SDS-PAGE)

form

solution

specific activity

>100,000 C5H50 units/mg protein

concentration

1 mg/mL in PBS, pH 7.2

technique(s)

activity assay: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... C5(727)

Application

Complement C5 is part of the terminal sequence in the complement pathway. In particular, it is cleaved into C5a and C5b which are responsible for chemotaxis, inflammation, and initiating the formation of the membrane attack complex (MAC). Drugs that inhibit complement C5 have been used in Paroxysmal nocturnal hemoglobinuria (PNH) patients to lessen thrombotic complications by lessening intravascular hemolysis.

Biochem/physiol Actions

Complement component C5 is processed by convertase enzymes in a cascade modulated in a unique way. The multi-subunit catalytic complex initially shows little activity against C5, but instead cleaves C3. A C3 cleavage product C3b covalently attaches to the complex and shifts its specificity to C5 by a factor of 1000.

Analysis Note

Functionally active by a sensitive hemolytic assay.

Other Notes

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library