Skip to Content
Merck

C3105

Coelenterazine fcp

≥95% (HPLC), solid

Synonym(s):

8-(cyclopentylmethyl)-2-[(4-fluorophenyl)methyl]-6-(4-hydroxyphenyl)-imidazo[1,2-a]pyrazin-3(7H)-one, CLZN-fcp

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H24FN3O2
CAS Number:
Molecular Weight:
417.48
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Coelenterazine fcp, solid

assay

≥95% (HPLC)

Quality Segment

form

solid

solubility

methanol: 1 mg/mL

storage temp.

−20°C

SMILES string

Oc1ccc(cc1)C2=CN3C(=O)C(Cc4ccc(F)cc4)=NC3=C(CC5CCCC5)N2

InChI

1S/C25H24FN3O2/c26-19-9-5-17(6-10-19)14-22-25(31)29-15-23(18-7-11-20(30)12-8-18)27-21(24(29)28-22)13-16-3-1-2-4-16/h5-12,15-16,27,30H,1-4,13-14H2

InChI key

OLXXULUJSSTXSP-UHFFFAOYSA-N

Biochem/physiol Actions

135 times higher luminescence intensity than native coelenterazine


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library