Skip to Content
Merck
All Photos(1)

Key Documents

C-26230

Sigma-Aldrich

Renal Epithelial Cell Growth Medium 2

Basal Medium, 500 ml

Synonym(s):

Recommended with C-39606 or C-39605

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.81

form

aqueous solution

packaging

pkg of 500 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Renal Epithelial Cell Basal Medium 2 (supplements sold separately)

To form a complete medium, it should be combined with the media supplements: C-39606 or C-39605

Application

Renal Epithelial Cell Growth Medium 2 is an optimized low-serum (5% V/V) medium that additionally contains Epinephrine. The combination of low serum and selected growth factors ensures successful growth in vitro making the Renal Epithelial Cell Growth Medium 2 the first choice for PromoCell Renal Epithelial Cells. The Renal Epithelial Cell Growth Medium 2 is optimized for primary human cells, but can also be used for porcine and mouse renal epithelial cells.Recommended for:Human Renal Epithelial Cells (HREpC)Human Renal Cortical Epithelial Cells (HRCEpC)

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials

Quality

All lots of PromoCell Renal Epithelial Cell Media are subjected to comprehensive quality control tests using primary human renal epithelial cells. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested renal epithelial cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Storage and Stability

Store the Basal Medium at 4 to 8°C in the dark immediately after arrival. Do not freeze the Basal Medium. If stored properly, the medium is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C

Subculture Routine

Click here for more information.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

PromoCell offers optimized primary human renal epithelial cells and growth medium with protocols for handling and cultivation.

PromoCell offers optimized primary human renal epithelial cells and growth medium with protocols for handling and cultivation.

PromoCell offers optimized primary human renal epithelial cells and growth medium with protocols for handling and cultivation.

PromoCell offers optimized primary human renal epithelial cells and growth medium with protocols for handling and cultivation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service